Learn More
anti-CD17, DyLight 755, Clone: MEM-68, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP150060IR
Catalog No. NBP150060IR
Add to cart
Description
Description
CD17 Monoclonal antibody specifically detects CD17 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD17
MEM-68
Flow Cytometry
Purified
Mouse
IgM
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Flow Cytometry
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
and amp;#223;-D-galactosyl-(1-4)- and amp;#223;-D-glucosyl-(1-1 and #039;)-ceramide, Lactosylceramide
Pre-B cell line NALM-6
Ion exchange chromatography
RUO
Primary
The mouse monoclonal antibody MEM-68 recognizes CD17, a membrane lipid moiety lactosylceramide expressed on granulocytes, monocytes and platelets.