Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD173 Antibody (BRIC231), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062120V
Description
CD173 Monoclonal specifically detects CD173 in Human samples. It is validated for Flow Cytometry.Specifications
CD173 | |
Monoclonal | |
DyLight 405 | |
H2, Type 2 H blood group antigen | |
HEL cell line | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Flow Cytometry | |
BRIC231 | |
Flow Cytometry | |
Mouse | |
Affinity Purified | |
RUO | |
NB100-62120 recognises the type 2 H blood group antigen, recently designated CD173. Active H substances in man, are expressed by many cells and tissues and also by erythrocytes | |
Store at 4C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction