Learn More
anti-CD19, DyLight 405, Clone: C19/366, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233194V
Catalog No. NBP233194V
Add to cart
Description
Description
CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications
Specifications
CD19
C19/366
Immunohistochemistry-Paraffin
Purified
CD19
A recominant protein fragment specific to CD19 was used as the immunogen for the C19/366 antibody.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human
Immunohistochemistry (Paraffin)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
B4, B-lymphocyte antigen CD19, B-lymphocyte surface antigen B4, CD19 antigen, CD19 molecule, CVID3, Differentiation antigen CD19, MGC12802, T-cell surface antigen Leu-12
Mouse
IgG1
0.1 ml
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers
Primary
930