Learn More
anti-CD21, Biotin, Clone: BB6-11C9.6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128247
Catalog No. NBP128247
Add to cart
Description
Description
CD21 Monoclonal antibody specifically detects CD21 in Mouse, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
Specifications
CD21
BB6-11C9.6
Biotin
PBS with 0.05% Sodium Azide
C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor
Mouse
IgG1 Kappa
0.25 mg
Dendritic Cell Markers
Primary
1380
Mouse, Porcine
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
CR2
Porcine CD21, Mr 160 kDa
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Porcine CD21, Mr 160 kDa