Learn More
anti-CD21, DyLight 680, Clone: BB6-11C9.6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128245FR
Catalog No. NBP128245FR
Add to cart
Description
Description
CD21 Monoclonal antibody specifically detects CD21 in Mouse, Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
CD21
BB6-11C9.6
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
CR2
Porcine CD21, Mr 160 kDa
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Porcine CD21, Mr 160 kDa
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
C3DRSLEB9, CD21, CD21 antigen, Complement C3d receptor, complement component (3d/Epstein Barr virus) receptor 2, complement receptor type 2, Cr2, EBV receptor, Epstein-Barr virus receptor
Mouse
IgG1 Kappa
0.25 ml
Adaptive Immunity, Dendritic Cell Markers, Immunology
Primary
1380
Mouse, Porcine