Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Mouse CD277/BTN3A1, DyLight 350, Clone: BT3.1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200227UV
Description
CD277/BTN3A1 Monoclonal specifically detects CD277/BTN3A1 in Human samples. It is validated for Flow Cytometry.Specifications
CD277/BTN3A1 | |
Monoclonal | |
DyLight 350 | |
butyrophilin, subfamily 3, member A1, dJ45P21.3 (butyrophilin, subfamily 3, member A1) | |
Mouse | |
Affinity Purified | |
RUO | |
11119 | |
Store at 4C in the dark. |
Flow Cytometry | |
BT3.1 | |
Flow Cytometry | |
BTN3A1 | |
The immunogen for this antibody is CD277 (B7 family, Butyrophilin). | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only