Learn More
anti-CD28, Clone: 204.12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142206
Catalog No. NBP142206
Add to cart
Description
Description
CD28 Monoclonal antibody specifically detects CD28 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, CyTOF-ready.
Specifications
Specifications
CD28
204.12
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, CyTOF-ready
Purified
CD28
The antibody reacts with the CD28 antigen.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
204-12 remained unassigned at the VI international workshop on human leucocyte differentiation antigens. Source: A BALB/c mouse was immunized with stimulated human leucocytes. Splenocytes were fused with NS1.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), CyTOF
Unconjugated
0.1 mg/ml PBS with 0.05% Sodium Azide
CD28 antigen, CD28 antigen (Tp44), CD28 molecule, MGC138290, T-cell-specific surface glycoprotein CD28, Tp44
Mouse
IgG2a
0.1 mg
Adaptive Immunity, Apoptosis, Immunology
Primary
940
Human, Mouse