Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD3, Clone: 4D10A6, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP147336

Catalog No. NBP147336


Add to cart

Description

Description

CD3 zeta Monoclonal antibody specifically detects CD3 zeta in Human samples. It is validated for Flow Cytometry, ELISA.
Specifications

Specifications

CD3 zeta
4D10A6
Flow Cytometry 1:200 - 1:1000, ELISA 1:10000
Affinity Purified
CD247
Purified recombinant fragment of human CD3 expressed in E. Coli.
Ascites
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute to desired concentration in diH2O.
ELISA, Flow Cytometry
Unconjugated
Ascites with 0.03% Sodium Azide
CD247 antigen, CD247 antigen, zeta subunit, CD247 molecule, CD3H, CD3Q, CD3z antigen, zeta polypeptide (TiT3 complex), CD3ZT-cell receptor T3 zeta chain, T3Z, T-cell antigen receptor complex, zeta subunit of CD3, T-cell surface glycoprotein CD3 zeta chain, TCRZCD3-ZETA
Mouse
IgG2a
0.1 ml
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology, Innate Immunity, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary
919
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only