Learn More
anti-CD3 epsilon, Biotin, Clone: BB23-8E6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128231
Catalog No. NBP128231
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD3 epsilon Monoclonal antibody specifically detects CD3 epsilon in Porcine samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
Specifications
CD3 epsilon
BB23-8E6
Biotin
PBS with 0.05% Sodium Azide
CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE
Mouse
IgG2b Kappa
0.25 mg
Adaptive Immunity, Apoptosis, Diabetes Research, Immunology, Innate Immunity, Signal Transduction
Primary
916
Porcine
Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen)
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
CD3E
Porcine CD3 epsilon, Mw 20 kDa
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Porcine CD3 epsilon, Mr 20 kDa