Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Rabbit anti-Hamster, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524411

Catalog No. PIPA524411


Add to cart

Description

Description

Cell adhesion molecule which is required for leukocyte transendothelial migration (TEM) under most inflammatory conditions. Tyr-679 plays a critical role in TEM and is required for efficient trafficking of PECAM1 to and from the lateral border recycling compartment (LBRC) and is also essential for the LBRC membrane to be targeted around migrating leukocytes. Prevents phagocyte ingestion of closely apposed viable cells by transmitting  ft.detachment ft. signals, and changes function on apoptosis, promoting tethering of dying cells to phagocytes (the encounter of a viable cell with a phagocyte via the homophilic interaction of PECAM1 on both cell surfaces leads to the viable cell ft.s active repulsion from the phagocyte. During apoptosis, the inside-out signaling of PECAM1 is somehow disabled so that the apoptotic cell does not actively reject the phagocyte anymore. The lack of this repulsion signal together with the interaction of the eat-me signals and their respective receptors causes the attachment of the apoptotic cell to the phagocyte, thus triggering the process of engulfment). Modulates BDKRB2 activation. Induces susceptibility to atherosclerosis.
Specifications

Specifications

CD31
Polyclonal
PBS with 0.09% sodium azide
Q08481
CD31 antigen, CD31/EndoCAM, GPIIA′, PECA1, PECAM-1, CD31/EndoCAM, adhesion molecule, endoCAM, platelet endothelial cell adhesion molecule
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100774872, 18613
Western Blot
Unconjugated
Pecam1
Liquid
PECAM1
KLH conjugated synthetic peptide between 508-536 amino acids from the C-terminal region of mouse CD31
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Hamster, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit