Learn More
anti-CD35, Alexa Fluor 700, Clone: E11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233080AF700
Catalog No. NBP233080Z
Add to cart
Description
Description
CD35 Monoclonal antibody specifically detects CD35 in Human, Primate samples. It is validated for ELISA.
Specifications
Specifications
CD35
E11
ELISA
Purified
CR1
Intact human monocytes were used as immunogen to generate the CD35 antibody (Hogg et al, 1984).
215 kDa
0.1 ml
Store at 4C in the dark.
Monoclonal
Recognizes a protein of 210-220kDa, which is identified as the complement receptor 1 (CR1)/CD35. This MAb is specific for a site in CR1 that is distal from the C3b-binding site, so that it is unable to block CR1 activity. This MAb is highly specific to CR1 and shows no cross-reaction with CR2. The primary function of CR1 is to serve as the cellular receptor for C3b and C4b, the most important components of the complement system leading to clearance of foreign macromolecules. The Knops blood group system is a system of antigens located on this protein. Follicular dendritic cells (FDC) are restricted to the B-cell regions of secondary lymphoid follicles. They are CD21+/CD35+/CD1a-. This MAb labels follicular dendritic cells and follicular dendritic cell sarcoma.
ELISA
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
complement component (3b/4b) receptor 1 (Knops blood group), KN
Mouse
IgG1 Kappa
Protein G purified
RUO
Primary
1378
Human, Primate