Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse CD35, PE, Clone: SPM554, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP234796PE

Catalog No. NBP234796PE


Add to cart

Description

Description

CD35 Monoclonal antibody specifically detects CD35 in Human, Baboon, Primate samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD35
SPM554
Flow Cytometry
Purified
CR1
Intact human monocytes
Protein A purified
RUO
Primary
1378
Baboon, Human, Primate
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
complement component (3b/4b) receptor 1 (Knops blood group), KN
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
Recognizes a protein of 210-220kDa, which is identified as the complement receptor 1 (CR1)/CD35. This MAb is specific for a site in CR1 that is distal from the C3b-binding site, so that it is unable to block CR1 activity. This MAb is highly specific to CR1 and shows no cross-reaction with CR2. The primary function of CR1 is to serve as the cellular receptor for C3b and C4b, the most important components of the complement system leading to clearance of foreign macromolecules. The Knops blood group system is a system of antigens located on this protein.Follicular dendritic cells (FDC) are restricted to the B-cell regions of secondary lymphoid follicles. They are CD21+/CD35+/CD1a-. This MAb labels follicular dendritic cells and follicular dendritic cell sarcoma.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only