Learn More
anti-CD40 Ligand/TNFSF5, Clone: TRAP1.3.6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142527
Catalog No. NBP142527
Add to cart
Description
Description
CD40 Ligand/TNFSF5 Monoclonal antibody specifically detects CD40 Ligand/TNFSF5 in Human, Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
Specifications
CD40 Ligand/TNFSF5
TRAP1.3.6
Unconjugated
PBS pH 7.4 with 0.09% Sodium Azide
CD154, CD154 antigen, CD40 antigen ligand, CD40 ligand, CD40-L, CD40LIGM, gp39, hCD40L, HIGM1, T-B cell-activating molecule, T-BAM, T-cell antigen Gp39, TNF-related activation protein, TNFSF5IMD3, TRAPtumor necrosis factor (ligand) superfamily, member 5 (hyper-IgM syndrome), tumor necrosis factor (ligand) superfamily member 5, Tumor necrosis factor ligand superfamily member 5
Mouse
IgG1
0.1 mg
Adaptive Immunity, Apoptosis, Asthma, Cancer, Immunology, Innate Immunity
Primary
959
Human, Mouse
Flow Cytometry, CyTOF
1.0 mg/mL
Flow Cytometry 1:10-1:1000, CyTOF-ready
Purified
CD40LG
Mouse myeloma cell line transfected with human CD40L (CD154)
Protein A or G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
TRAP1.3.6 recognises the human CD154 cell surface antigen, a 32kD glycoprotein also known as CD40 ligand. CD154 is expressed on activated T lymphocytes, predominantly CD4 +ve and also on some basophils and mast cells. TRAP1.3.6 binds to CD154 at an epitope distinct from the CD40 binding site. Immunogen: Mouse myeloma cell line transfected with human CD40L (CD154)