Learn More
anti-CD43/Sialophorin, DyLight 755, Clone: DF-T1, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233140IR
Catalog No. NBP233140IR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD43/Sialophorin
DF-T1
ELISA
Purified
SPN
Myeloblastic KG1 cells were used as the immunogen for this antibody.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
It recognizes a cell surface glycoprotein of 95/115/135kDa (depending upon the extent of glycosylation), identified as CD43. 70-90% of T-cell lymphomas and from 22-37% of B-cell lymphomas express CD43. No reactivity has been observed with reactive B-cells. So a B-lineage population that co-expresses CD43 is highly likely to be a malignant lymphoma, especially a low-grade lymphoma, rather than a reactive B-cell population. When CD43 antibody is used in combination with anti-CD20, effective immunophenotyping of the lymphomas in formalin-fixed tissues can be obtained. Co-staining of a lymphoid infiltrate with anti-CD20 and anti-CD43 argues against a reactive process and favors a diagnosis of lymphoma.
ELISA
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
CD43 antigen, CD43), Galactoglycoprotein, GALGP, Leukocyte sialoglycoprotein, Sialophorin, sialophorin (gpL115, leukosialin, CD43)
Mouse
IgG1 Kappa
0.1 ml
Immunology
Primary
6693
Human