Learn More
anti-CD44, FITC, Clone: AV6, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128346
Catalog No. NBP128346
Add to cart
Description
Description
CD44 Monoclonal antibody specifically detects CD44 in Chicken, Turkey samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
Specifications
CD44
AV6
FITC
PBS with 0.05% Sodium Azide
CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2CD44 antigen (homing function and Indian blood group system), MDU3CDW44, MIC4MGC10468, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I
Mouse
IgG1 Kappa
0.25 mg
Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
960
Chicken, Turkey
Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Frozen)
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
CD44
Chicken CD44 (Mr 90-kDa).
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Chicken CD44 (Mr 90-kDa).