Learn More
anti-CD45, Biotin, Clone: LT40, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128297
Catalog No. NBP128297
Add to cart
Description
Description
CD45 Monoclonal antibody specifically detects CD45 in Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
CD45
LT40
Biotin
PBS with 0.05% Sodium Azide
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide
Mouse
IgM Kappa
0.25 mg
Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary
5788
Chicken
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 units, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
PTPRC
Chicken bursal lymphocytes
IgM purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Chicken CD45 (Mr 190 to 215-kDa).