Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD45 Antibody (LT40), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128296
Description
CD45 Monoclonal specifically detects CD45 in Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Product Image.Specifications
CD45 | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
PTPRC | |
Chicken bursal lymphocytes | |
0.25 mg | |
Innate Immunity, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
5788 | |
Chicken | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
LT40 | |
FITC | |
B220, CD45 antigen, CD45R, EC 3.1.3.48, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
Mouse | |
IgM purified | |
RUO | |
Primary | |
Chicken CD45 (Mr 190 to 215-kDa). | |
Store at 4C in the dark. | |
IgM κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction