Learn More
anti-CD46, Clone: M177, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP151131
Catalog No. NBP151131
Add to cart
Description
Description
CD46 Monoclonal antibody specifically detects CD46 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry-Frozen, Functional, Immunoassay.
Specifications
Specifications
CD46
M177
Unconjugated
0.2 um filtered solution in PBS and 0.1% BSA with No Preservative
AHUS2, CD46 antigen, CD46 molecule, complement regulatory protein, membrane cofactor protein (CD46, trophoblast-lymphocyte cross-reactive antigen), MIC10, TLXcomplement regulatory protein, trophoblast leucocyte common antigen, Trophoblast leukocyte common antigen, trophoblast-lymphocyte cross-reactive antigen
Mouse
IgG1
0.05 mg
Cancer, Cell Biology, Cellular Markers, Immunology
Primary
4179
Flow Cytometry, Functional Assay, Immunoassay, Immunoprecipitation, Western Blot
0.1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Functional, Immunoassay
Purified
CD46
Purified MCP
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human