Learn More
anti-CD47, Clone: BRIC126, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142488
Catalog No. NBP142488
Add to cart
Description
Description
CD47 Monoclonal specifically detects CD47 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
Specifications
CD47
BRIC126
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
CD47
Human erythrocytes.
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Recognizes the human CD47 cell surface glycoprotein, a heavily N-glycosylated 47-52kD molecule. CD47 is expressed on all cells and tissues so far examined, although expression is reduced on erythrocytes of the rare Rh null phenotype.
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Unconjugated
Tris-Glycerine (pH 7.8) with 0.09% Sodium Azide
antigen identified by monoclonal 1D8, Antigenic surface determinant protein OA3, CD47 antigen, CD47 antigen (Rh-related antigen, integrin-associated signal transducer), CD47 glycoprotein, CD47 molecule, IAPintegrin associated protein, Integrin-associated protein, leukocyte surface antigen CD47, MER6integrin-associated signal transducer, OA3, Protein MER6, Rh-related antigen
Mouse
IgG2b
0.1 mg
Immunology, Signal Transduction
Primary
961
Human