Learn More
anti-CD5, Biotin, Clone: 2-191, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP128317
Catalog No. NBP128317
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD5
2-191
Biotin
PBS with 0.05% Sodium Azide
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
Mouse
IgG1 Kappa
0.25 mg
Store at 4C. Do not freeze.
Monoclonal
Chicken CD5, Mr 64 kDa (protein core of 57 kDa)
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunoprecipitation
0.5mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunocytochemistry
Purified
CD5
Embryonic day 15 thymocytes
Protein A or G purified
RUO
Primary
921
Chicken