Learn More
anti-CD5, Clone: 2-191, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP128315
Catalog No. NBP128315
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CD5
2-191
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
Mouse
IgG1 Kappa
0.5 mg
Store at 4C. Do not freeze.
Monoclonal
Chicken CD5, Mr 64 kDa (protein core of 57 kDa)
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Immunocytochemistry
0.5 mg/mL
Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Immunocytochemistry
Purified
CD5
Embryonic day 15 thymocytes
Protein A or G purified
RUO
Primary
921
Chicken