Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD5, Clone: 2-4-2B1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142353

Catalog No. NBP142353


Add to cart

Description

Description

CD5 Monoclonal antibody specifically detects CD5 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications

Specifications

CD5
2-4-2B1
Unconjugated
PBS and 50% Glycerol with 0.01% Sodium Azide
CD5 antigen, CD5 antigen (p56-62), CD5 molecule, LEU1T-cell surface glycoprotein CD5, Lymphocyte antigen T1/Leu-1, T1
Mouse
IgG1
0.2 ml
Adaptive Immunity, Cell Biology, Cytokine Research, Immunology
Primary
921
Rat
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
0.1 mg/mL
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
CD5
The antibody reacts with the CD5-antigen antigen.
Protein A or G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
This monoclonal antibody recognizes rat CD5 antigen (MW 69 kDa) and is reactive with all common inbred strains. 2-4-2B1 is derived from hybridization of mouse SP2/0 myeloma cells with spleen cells from Balb/c mice immunized with WAG/Rij spleen cells.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only