Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD72.1 Antibody (10-1.D.2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128023
Description
CD72.1 Monoclonal specifically detects CD72.1 in Mouse samples. It is validated for Flow Cytometry, In vitro assay.Specifications
CD72.1 | |
Monoclonal | |
0.5mg/mL | |
Flow Cytometry <= 1 ug/10^6 cells, In vitro assay | |
Cd72 | |
DBA/2 mouse spleen cells | |
0.25 mg | |
Primary | |
Mouse CD72.1/Lyb-2.1, 45 kDa | |
Store at 4C. Do not freeze. | |
IgG2b κ |
Flow Cytometry, In vitro Assay | |
10-1.D.2 | |
FITC | |
CD72 antigen, Ly19, Ly32, Lyb2, Lym19 | |
Mouse | |
Protein A or G purified | |
RUO | |
12517 | |
Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction