Learn More
anti-CD79B, Clone: CB3-1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128528
Catalog No. NBP128528
Add to cart
Description
Description
CD79B Monoclonal antibody specifically detects CD79B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
Specifications
CD79B
CB3-1
Unconjugated
BBS (pH 8.2) with No Preservative
B29B-cell-specific glycoprotein B29, B-cell antigen receptor complex-associated protein beta chain, CD79b antigen, CD79B antigen (immunoglobulin-associated beta), CD79b molecule, immunoglobulin-associated beta, IGBAGM6, Ig-beta, Immunoglobulin-associated B29 protein
Mouse
IgG1
0.1 mg
Store at 4C. Do not freeze.
Monoclonal
Human CD79 beta/B29/Ig beta
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
0.1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry <= 1 ug/10^6 cells, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500
Purified
CD79B
Ramos B cells
Protein A or G purified
RUO
Primary
974
Human