Learn More
CD8 Mouse anti-Porcine, Clone: 76-2-11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128237
Catalog No. NBP128237
Add to cart
Description
Description
CD8 Monoclonal antibody specifically detects CD8 in Porcine samples. It is validated for Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Block/Neutralize.
Specifications
Specifications
CD8
76-2-11
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
CD8, CD8a molecule, LEU2, p32
Mouse
IgG2b Kappa
0.25 mg
Innate Immunity, Stem Cell Markers
Primary
925
Porcine
Blocking Assay, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Blocking/Neutralizing 50-200 molar excess
Purified
CD8A
Fresh dd miniature swine thymocytes
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Porcine CD8a, Mr 32-34 kDa