Learn More
CD8 Mouse anti-Avian, Chicken, Turkey, Clone: CT-8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128270
Catalog No. NBP128270
Add to cart
Description
Description
CD8 Monoclonal antibody specifically detects CD8 in Avian, Chicken, Turkey samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry, Block/Neutralize.
Specifications
Specifications
CD8
CT-8
Unconjugated
0.1M BBS (pH 8.2) with No Preservative
CD8, CD8a molecule, LEU2, p32
Mouse
IgG1 Kappa
0.5 mg
Innate Immunity, Stem Cell Markers
Primary
925
Avian, Chicken, Turkey
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Blocking/Neutralizing 50-200 molar excess, Immunocytochemistry
Purified
CD8A
Chicken thymocytes and Ig-negative blood leukocytes
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
alpha chain (34-kDa) of chicken CD8