Learn More
CD8 Mouse anti-Porcine, DyLight 680, Clone: 76-2-11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128237FR
Catalog No. NBP128237FR
Add to cart
Description
Description
CD8 Monoclonal antibody specifically detects CD8 in Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
Specifications
CD8
76-2-11
Flow Cytometry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Purified
CD8A
Fresh dd miniature swine thymocytes
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Porcine CD8a, Mr 32-34 kDa
Flow Cytometry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
CD8, CD8a molecule, LEU2, p32
Mouse
IgG2b Kappa
0.25 ml
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary
925
Porcine