Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Mouse anti-Porcine, PE, Clone: 76-2-11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP128237PE

Catalog No. NBP128237PE


Add to cart

Description

Description

CD8 Monoclonal antibody specifically detects CD8 in Porcine samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD8
76-2-11
Flow Cytometry
Purified
CD8A
Fresh dd miniature swine thymocytes
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Porcine CD8a, Mr 32-34 kDa
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
CD8, CD8a molecule, LEU2, p32
Mouse
IgG2b Kappa
0.25 ml
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers
Primary
925
Porcine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only