Learn More
anti-CD8 beta, DyLight 350, Clone: EP42, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128300UV
Catalog No. NBP128300UV
Add to cart
Description
Description
CD8 beta Monoclonal antibody specifically detects CD8 beta in Chicken samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
CD8 beta
EP42
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Purified
CD8B
Chicken splenocytes
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
beta chain (Mr 34 kDa) of chicken CD8
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
CD8 antigen, beta polypeptide 1 (p37), CD8b antigen, CD8b molecule, CD8B1P37, LEU2, LY3, LYT3, MGC119115, T lymphocyte surface glycoprotein beta chain, T-cell surface glycoprotein CD8 beta chain
Mouse
IgG2a Kappa
0.25 ml
Adaptive Immunity, Cytokine Research, Immunology, Innate Immunity, Signal Transduction, Stem Cell Markers
Primary
926
Chicken