Learn More
CD9 Mouse anti-Human, Canine, Feline, Hamster, Monkey, Rabbit, PE, Clone: MM2/57, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP128365
Catalog No. NBP128365
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD9 Monoclonal antibody specifically detects CD9 in Human, Canine, Feline, Hamster, Monkey, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen).
Specifications
Specifications
CD9
MM2/57
PE
PBS with Sodium Azide
Purified
CD9
The immunogen is a peptide made to the amino acid region CD9
Protein A or G purified
RUO
Primary
928
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
0.1 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 10 uL/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
P21926
BA2, BA-2/p24 antigen, BTCC-1, CD9 antigen, CD9 molecule, Cell growth-inhibiting gene 2 protein, DRAP-27, Leukocyte antigen MIC3, motility related protein-1,5H9 antigen, Motility-related protein, MRP-1FLJ99568, P24, Tetraspanin-29, TSPAN-29, TSPAN29MIC3CD9 antigen (p24)
Mouse
IgG2b Kappa
1 ml
Store at 4C. Do not freeze.
Monoclonal
Human, Canine, Feline, Hamster, Monkey, Rabbit