Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
anti-CEACAM-1/CD66a, DyLight 488, Clone: COL-1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233306G
Catalog No. NBP233306G
Description
Description
CEACAM/CD66 Monoclonal antibody specifically detects CEACAM/CD66 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
CEACAM/CD66
COL-1
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1-5ug/ml for coating, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
Mouse
IgG2a Kappa
0.1 ml
Apoptosis, Cancer
Primary
1048
Human
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
CEACAM1
Colon carcinoma cell extract was used as the immunogen for the CEACAM1 antibody.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This antibody recognizes proteins of 80-200kDa, identified as different members of CEACAM family. This MAb does not react with nonspecific cross-reacting antigen (NCA) and with human polymorphonuclear leucocytes. It shows no reaction with a variety of normal tissues and is suitable for staining of formalin/paraffin tissues. CEACAM is not found in benign glands, stroma, or malignant prostatic cells