Learn More
anti-ChABC, Alexa Fluor 488, Clone: 1E10, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196141AF488
Catalog No. NBP196141X
Add to cart
Description
Description
ChABC Monoclonal antibody specifically detects ChABC in Rat, Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
ChABC
10000000000
Western Blot, ELISA
Purified
Mouse
IgG
Protein G purified
RUO
Primary
Non-species specific
ELISA, Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Chondroitinase ABC
Chondroitinase ABC
80 kDa
0.1 ml
Store at 4C in the dark.
Monoclonal