Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
anti-ChABC, Alexa Fluor 488, Clone: 6A12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196142AF488
Catalog No. NBP196142X
Description
Description
ChABC Monoclonal antibody specifically detects ChABC in Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
ChABC
6A12
Western Blot, ELISA
Purified
Mouse
IgG
Protein G purified
RUO
Primary
Non-species specific
ELISA, Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
Chondroitinase ABC
Chondroitinase ABC
80 kDa
0.1 ml
Store at 4C in the dark.
Monoclonal