Learn More
anti-ChABC, DyLight 405, Clone: 6A12, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196142V
Catalog No. NBP196142V
Add to cart
Description
Description
ChABC Monoclonal antibody specifically detects ChABC in Non-species specific samples. It is validated for Western Blot, ELISA.
Specifications
Specifications
ChABC
6A12
Western Blot, ELISA
Purified
Mouse
IgG
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
ELISA, Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Chondroitinase ABC
Chondroitinase ABC
80 kDa
0.1 ml
Neurodegeneration, Neuroscience
Primary
Non-species specific