Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-Cofilin, Clone: 1C1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP121044
Description
Cofilin Monoclonal specifically detects Cofilin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Cofilin | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:1000-1:2000, Flow Cytometry, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
18 kDa phosphoprotein, CFL, cofilin 1 (non-muscle), Cofilin, non-muscle isoform, cofilin-1, p18 | |
Mouse | |
Affinity Purified | |
RUO | |
1072 | |
Human | |
IgG1 κ |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
1C1 | |
Unconjugated | |
NP_005498 | |
CFL1 | |
Recombinant human Cofilin 1 (1-166aa) purified from E. coli | |
0.1 mL | |
Primary | |
Anti-human Cofilin 1 mAb, clone 1C1, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human Cofilin 1 protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only