Learn More
anti-Creatine Kinase BB, Clone: 893CT29.1.1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232292
Catalog No. NBP232292
Add to cart
Description
Description
Creatine Kinase BB Monoclonal antibody specifically detects Creatine Kinase BB in Human, Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Creatine Kinase B
893CT29.1.1
Western Blot 1:2000
P12277, P12277
B-CK, Creatine kinase B chain, creatine kinase, brain, creatine kinase-B, EC 2.7.3, EC 2.7.3.2
Mouse
IgG1
Unpurified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse
Western Blot
Unconjugated
Crude ascites with 0.09% Sodium Azide
Ascites
CKB
Purified His-tagged CKB protein was used to produced this monoclonal antibody.
42.644 kDa
100 ul
Cancer
Primary
1152