Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse CtIP, Alexa Fluor 488, Clone: 19E8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP141205AF488

Catalog No. NBP141205X


Add to cart

Description

Description

CtIP Monoclonal antibody specifically detects CtIP in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications

Specifications

CtIP
1900000000
Western Blot, Chromatin Immunoprecipitation, Immunocytochemistry/Immunofluorescence, Immunoprecipitation
Purified
RBBP8
Human CtIP
Protein G purified
RUO
Primary
5932
Human, Mouse
ChIP assay, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
CtBP-interacting protein, CtIP, DNA endonuclease RBBP8, EC 3.1, RBBP-8, retinoblastoma binding protein 8, Retinoblastoma-binding protein 8, Retinoblastoma-interacting protein and myosin-like, RIMSAE2, Sporulation in the absence of SPO11 protein 2 homolog
Mouse
IgG
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody will recognize human CtIP.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only