Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse CX3CR1, PE, Polyclonal, R&D Systems™

Manufacturer:  R&D Systems FAB5825P100

 View more versions of this product

Catalog No. FAB5825P100


Add to cart

Description

Description

Host Species: Goat
Isotype: IgG
Species Reactivity: Ms
Immunogen: E. coli-derived recombinant mouse CX3CR1. Met1-Thr32, Leu92-Lys104, Thr169-Val196, Lys258-Leu274

Applications:

Flow Cytometry

Specifications

Specifications

CX3CR1
R-PE
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Affinity Purified
CX3CR1
E. coli-derived recombinant mouse CX3CR1 Met1-Thr32, Leu92-Lys104, Thr169-Val196, Lys258-Leu274 Accession # Q9Z0D9
Antigen Affinity-purified
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Polyclonal
Detects mouse CX3CR1 in direct ELISAs. Stains mouse CX3CR1 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
Flow Cytometry 10 uL/10^6 cells
Q9Z0D9
Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKbRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28, V28C-X3-C CKR-1
Goat
IgG
100 Tests
Primary
1524
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit