Learn More
anti-Mouse Cytochrome c, Clone: 6H2, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP143254
Catalog No. NBP143254
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Cytochrome c
6H2
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
CYCHCS, cytochrome c, cytochrome c, somatic, THC4
Mouse
IgG1 Kappa
0.1 mg
Apoptosis, Cellular Markers, Cholesterol Metabolism, Core ESC Like Genes, Lipid and Metabolism, Mitochondrial Markers, Stem Cell Markers
Primary
54205
Flow Cytometry, Immunoprecipitation
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Flow (Intracellular)
Affinity Purified
CYCS
The immunogen for this antibody was Cytochrome c.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human, Mouse, Rat