Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse Cytokeratin 14, Clone: DE-SPK14, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP142262

Catalog No. NBP142262


Add to cart

Description

Description

Cytokeratin 14 Monoclonal antibody specifically detects Cytokeratin 14 in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Cytokeratin 14
DE-SPK14
Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Supernatant
KRT14
The antibody reacts with the Keratin 14 antigen.
Tissue culture supernatant
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
DE-SPK14 reacts specifically in immunoblotting with human keratin 14 (50 kD). In tissue sections this mAb provides a positive reaction on basal cells of non-keratinizing stratified epithelia, basal cells and suprabasal cells of the epidermis and gingival, myoepithelial cells and squamous cell carcinomas.
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Unconjugated
Tissue culture supernatant with No Preservative
CK14, CK-14, cytokeratin 14, cytokeratin-14, EBS3, EBS4, K14, keratin 14, keratin 14 (epidermolysis bullosa simplex, Dowling-Meara, Koebner), keratin, type I cytoskeletal 14, Keratin-14, NFJ
Mouse
IgG2b
0.2 ml
Cytoskeleton Markers
Primary
3861
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only