Learn More
anti-Mouse Cytokeratin 17, Clone: Ks 17.E3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP142265
Catalog No. NBP142265
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Cytokeratin 17 Monoclonal antibody specifically detects Cytokeratin 17 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Cytokeratin 17
Ks 17.E3
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
KRT17
The antibody reacts with the Keratin 17 antigen.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
This antibody represents an excellent marker to distinguish myoepithelial cells from luminal epithelium of various glands (mammary, sweat, salivary, bronchial, tracheal, laryngeal, esophageal) and benign from malignant forms of e.g. mammary gland tumors. Most malignant forms of mammary tumors are negative for cytokeratin 17. Antigen: Human cytokeratin 17 of Mr 46,000 Antigen recognized in species: Human and rat (Mr 40,000 polypeptide)
Human, Rat
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Unconjugated
Distiled water with No Preservative
39.1, CK-17, cytokeratin-17, K17, keratin 17, keratin, type I cytoskeletal 17, keratin-17, PC, PC2, PCHC1
Mouse
IgG2b
0.2 ml
Cancer
Primary
3872
Reconstitute with 1.0 ml sterilized distilled water to desired concentration.