Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse Cytokeratin 19, Clone: RCK108, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP142238

Catalog No. NBP142238

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Cytokeratin 19
RCK108
Western Blot 1:500-1:1000, Flow Cytometry 1:10-1:20, Immunohistochemistry 1:10-1:100, Immunocytochemistry/Immunofluorescence 1:10-1:100, Immunohistochemistry-Paraffin 1:10-1:100, Immunohistochemistry-Frozen 1:10-1:100
P08727
CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366
Mouse
IgG1
1 ml
Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers
Primary
3880
Human, Rat, Zebrafish
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Tissue culture supernatant with 0.09% Sodium Azide
Supernatant
KRT19
The antibody reacts with the Keratin 19 antigen.
Tissue culture supernatant
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
The monoclonal antibody specifically reacts with the cytokeratin 19, a 40 kDa polypeptide, which is found in most simple and non-keratinized stratified epithelia. This antibody reacts on formalin fixed, paraffin embedded material and can be used in diagnostic immunopathology. The reaction pattern of cytokeratin 19 is documented in the literature. Antigen origin: human bladder cell line T24. Antigen location: Cytoplasm.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only