Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse Cytokeratin, LMW, APC, Clone: AE1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP233178APC

Catalog No. NBP233178J


Add to cart

Description

Description

Cytokeratin, LMW Monoclonal antibody specifically detects Cytokeratin, LMW in Human, Mouse, Rat, Bovine, Canine, Chicken, Primate, Rabbit samples. It is validated for Flow Cytometry.
Specifications

Specifications

Cytokeratin, LMW
AE1
Flow Cytometry
Purified
KRT77
Human epidermal keratin was used as the immunogen for the Cytokeratin LMW AE1 antibody.
Protein G purified
RUO
Primary
374454
Human, Mouse, Rat, Bovine, Canine, Chicken, Other, Primate, Rabbit
Flow Cytometry
APC
PBS with 0.05% Sodium Azide
Cytokeratin LMW, cytokeratin-1B, keratin 77, keratin, type II cytoskeletal 1b, KRT77, type-II keratin Kb39
Mouse
IgG1 Kappa
0.1 ml
Store at 4C in the dark.
Monoclonal
This MAb recognizes the 56.5kDa (CK10); 50kDa (CK14); 50kDa (CK15); 48kDa (CK16); 40kDa (CK19) keratins of the acidic (Type I or LMW) subfamily. Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI less than 5.7) and basic (pI less than 6.0) subfamilies. The acidic keratins have molecular weights (MW) of 56.5, 55, 51, 50, 50', 48, 46, 45, and 40kDa. MAb AE3 recognizes the 65-67, 64, 59, 58, 56, and 52kDa keratins of basic subfamily. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. AE1/AE3 is a broad spectrum anti pan-keratin antibody cocktail, which differentiates epithelial tumors from non-epithelial tumors e.g. squamous vs. adenocarcinoma of the lung, liver carcinoma, breast cancer, and esophageal cancer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only