Learn More
Cytokeratin, pan Antibody (SPM115 + SPM116), HRP, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP234394H
Catalog No. NBP234394H
Add to Cart
Description
Description
Cytokeratin, pan Monoclonal specifically detects Cytokeratin, pan in Human, Mouse, Rat, Bovine, Canine, Chicken, Monkey, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
Cytokeratin, pan
Monoclonal
HRP
KRT1
This Cytokeratin, pan Antibody (SPM115 + SPM116) was developed against human epidermal keratin.
Protein A or G purified
RUO
Primary
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. This antibody cocktail recognizes acidic (Type I or LMW) and basic (Type II or HMW) cytokeratins, which 67kDa (CK1); 64kDa (CK3); 59kDa (CK4); 58kDa (CK5); 56kDa (CK6); 52kDa (CK8); 56.5kDa (CK10); 50kDa (CK14); 50kDa (CK15); 48kDa (CK16); 40kDa (CK19). Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. AE-1/AE-3 is a broad spectrum anti pan-cytokeratin antibody cocktail, which differentiates epithelial tumors from non-epithelial tumors e.g. squamous vs. adenocarcinoma of the lung, liver carcinoma, breast cancer, and esophageal cancer. It has been used to characterize the source of various neoplasms and to study the distribution of cytokeratin containing cells in epithelia during normal development and during the development of epithelial neoplasms. This antibody stains cytokeratins present in normal and abnormal human tissues
Store at 4C in the dark.
IgG1 κ
Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
SPM115 + SPM116
Western Blot, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Mouse
50 kDa
0.1 mL
Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction
3848
Human, Mouse, Rat, Bovine, Canine, Chicken, Primate, Rabbit
Purified
Product Suggestions
Product Suggestions