Learn More
anti-Mouse Daxx, Alexa Fluor 488, Clone: DAXX-03, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP144892AF488
Catalog No. NBP144892X
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Daxx
DAXX-03
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
DAXX
His6-tagged protein corresponding to amino acids 580-740 of human Daxx.
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
The antibody DAXX-03 reacts with C-terminal part of Daxx, a 110 kDa intracellular protein.
Western Blot, Immunocytochemistry, Immunofluorescence
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246
Mouse
IgG1
0.1 ml
Apoptosis, Cancer, Phospho Specific, Tumor Suppressors
Primary
1616
Human