Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse Daxx, Clone: 7A11, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP147453

Catalog No. NBP147453

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Daxx
7A11
Western Blot 1: 500-1:2000, Flow Cytometry, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000
Affinity Purified
DAXX
Purified recombinant fragment of human DAXX expressed in E. Coli.
81 kDa
0.1 ml
Apoptosis, Cancer, Tumor Suppressors
Primary
1616
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
PBS with 50% glycerol
BING2, CENP-C binding protein, DAP6MGC126245, Daxx, death domain-associated protein 6, death-associated protein 6, death-domain associated protein, EAP1, ETS1-associated protein 1, Fas death domain-associated protein, Fas-binding protein, hDaxx, MGC126246
Mouse
IgG1
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only