Learn More
anti-Mouse DCIR/CLEC4A, Alexa Fluor 647, Clone: 111F8.04, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals DDX0180A647100
Catalog No. DDX0180A647
Add to cart
Description
Description
DCIR/CLEC4A Monoclonal antibody specifically detects DCIR/CLEC4A in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
DCIR/CLEC4A
111F8.04
Alexa Fluor 647
PBS and 50% Glycerol with No Preservative
CLECSF6C-type lectin superfamily member 6, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 6, C-type lectin domain family 4 member A, C-type lectin domain family 4, member A, DCIRLLIR, DDB27C-type lectin DDB27, Dendritic cell immunoreceptor, Lectin-like immunoreceptor
Mouse
IgG1
0.1 mg
Immunology, Innate Immunity
Primary
50856
Flow Cytometry
0.5mg/mL
Flow Cytometry 1:10-1:1000
Ion Exchange Chromatography
CLEC4A
DCIR was used as immunogen.
Ion exchange chromatography
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Human