Learn More
anti-Mouse Defensin alpha 5, DyLight 405, Clone: 8C8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB11060002V
Catalog No. NB11060002V
Add to cart
Description
Description
Defensin alpha 5 Monoclonal specifically detects Defensin alpha 5 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
Defensin alpha 5
8C8
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Q01523
DEF5defensin, alpha 5, preproprotein, defensin 5, Defensin, alpha 5, defensin, alpha 5, Paneth cell-specific, defensin-5, HD-5, MGC129728
Mouse
IgG2b
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
This is specific for HD5. It does not cross-react with HD6, lysozyme or sPLA2.
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Purified
DEFA5
Full-length human Defensin alpha 5 [Swiss-Prot# Q01523]. The propeptide is the full-length protein. In the mature form, it is cleaved at residue 63 (alanine).
10 kDa
0.1 ml
Immunology, Innate Immunity
Primary
1670
Human