Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Desmoglein-3, Clone: 4W25, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236422

Catalog No. NBP236422

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Desmoglein-3
4W25
Unconjugated
PBS with No Preservative
Cadherin family member 6, CDHF6130-kD pemphigus vulgaris antigen, desmoglein 3, desmoglein 3 (pemphigus vulgaris antigen), desmoglein-3, DKFZp686P23184,130 kDa pemphigus vulgaris antigen, pemphigus vulgaris antigen, PVA
Mouse
IgG1
0.1 mg
Cytoskeleton Markers, Extracellular Matrix, Immune System Diseases, Signal Transduction
Primary
1830
Human
Immunohistochemistry (Paraffin), Western Blot
LYOPH
Western Blot 1:100 - 1:2000 Immunohistochemistry-Paraffin 1:10 - 1:500
Purified
DSG3
recombinant human Desmoglein-3
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only