Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Mouse DFF45/ICAD, Clone: 5C4, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP147724

Catalog No. NBP147724

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

DFF45/ICAD
OTI5C4
Unconjugated
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
DFF1DNA fragmentation factor 45 kDa subunit, DFF45DNA fragmentation factor subunit alpha, DFF-45DNA fragmentation factor, 45 kD, alpha polypeptide, DNA fragmentation factor, 45kDa, alpha polypeptide, ICAD, Inhibitor of CAD
Mouse
IgG1
0.1 ml
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Tumor Suppressors
Primary
1676
Human
Immunocytochemistry, Immunofluorescence, Western Blot
2.0mg/mL
Western Blot 1:2500-5000, Immunocytochemistry/Immunofluorescence 1:50-100
Affinity Purified
DFFA
Recombinant protein expressed in E.coli corresponding to amino acids 1-331 of human DFF45
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
This antibody is specific for Homo sapiens DNA fragmentation factor, 45kDa, alpha polypeptide (DFFA), transcript variant 1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only